Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Cyclohexanone, Supelco®

Cyclohexanone, Supelco®

Glass bottle, 1 mL
Catalog # 02482-1ML
Flammable
Irritant
Supplier:   Merck
Cyclohexanone, Supelco®
Catalog # 02482-1ML
Supplier:  Merck
CAS Number:  108-94-1
Cyclohexanone, Supelco®
Cyclohexanone, Supelco®

Glass bottle, 1 mL

CAS Number:  108-94-1
Price
SFr. 46.30
Log in for availability
1 * 1 mL
Product Details & Documents
Synonyms:  Pimelic Ketone
  • CAS Number:108-94-1
  • Formula:C₆H₁₀(=O)
  • MDL Number:MFCD00001625
  • Molecular Weight:98.14 g/mol
  • Merck Index:13,02755
  • Boiling Pt:155.6 °C (1013 hPa)
  • Melting Pt:–31 °C
  • Density:0.9478 g/cm³ (20 °C)
  • Flash Pt:43 °C
  • UN:1915
  • ADR:3,III
  •  :M-Clarity™ quality level = MQ100
  • Storage Temperature:Ambient
  • EINECS:203-631-1
  • REACH:01-2119453616-35

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.