Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Cyclohexanone ≥99%

Cyclohexanone ≥99%

Flammable
Irritant
   Sustainable Options Available
Cyclohexanone ≥99%
   Sustainable Options Available
Certificates
Cyclohexanone ≥99%

2 Options Available Below

Product Details & Documents

Purity: 99+% CAS No.: 108-94-1 Molecular Formula: C6H10O Formula Weight: 98.15 Flash Point: 47°(116°F) Refractive Index: 1.4507 Melting Point: -26° Boiling Point: 154-156° Beilstein Reference: 385735 Merck Reference: 14,2726 UN No.: 1915 MDL No.: MFCD00001625

Synonyms:  Pimelic Ketone
  • CAS Number:108-94-1
  • Formula:C₆H₁₀(=O)
  • MDL Number:MFCD00001625
  • Molecular Weight:98.14 g/mol
  • Merck Index:14,2726
  • Boiling Pt:154…156 °C
  • Melting Pt:–26 °C
  • Density:0.948
  • Flash Pt:47 °C (116 °F)
  • Storage Temperature:Ambient
  • EINECS:203-631-1
  • UN:1915
  • ADR:3,III
  • REACH:01-2119453616-35
Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • A15607.AP
  • A15607.0F
  • Environmentally Preferable:
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Environmental certification:
  • Chem21 Solvent Selection Guide recommended by default
  • Chem21 Solvent Selection Guide recommended by default

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - SFr. 68.80
    SFr. 68.80
    1 * 500 mL
       SFr. 68.80

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    A15607.AP
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended by default
      Safer Human and Environmental Health
  • Glass bottle
    2.500 mL
    Log in for availability
    Log in for availability
    1 * 2.500 mL
    1 * 2.500 mL - SFr. 164.00
    SFr. 164.00
    1 * 2.500 mL
       SFr. 164.00

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    A15607.0F
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended by default
      Safer Human and Environmental Health