Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Cyclohexanone 99.8%, extra pure

Irritant
Corrosive
Flammable
   Sustainable Options Available
Cyclohexanone 99.8%, extra pure
   Sustainable Options Available
Certificates
Cyclohexanone 99.8%, extra pure

4 Options Available Below

Product Details & Documents
Synonyms:  Pimelic Ketone
  • CAS Number:108-94-1
  • Formula:C₆H₁₀(=O)
  • MDL Number:MFCD00001625
  • Molecular Weight:98.14 g/mol
  • Merck Index:13,02755
  • Boiling Pt:155 °C (760 mmHg)
  • Melting Pt:–47 °C
  • Density:0.947 g/cm³
  • Flash Pt:43 °C
  • Storage Temperature:Ambient
  • EINECS:203-631-1
  • UN:1915
  • ADR:3,III
  • REACH:01-2119453616-35
Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4450 to 1.4520 (20°C, 589 nm)
  • Water:
    ≤0.2 % (K.F.)
  • GC:
    ≥99.75 %
  • Color scale:
    ≤20 APHA
  • Iron (Fe):
    ≤0.001 %
  • Magnesium (Mg):
    ≤0.005 %
  • Manganese (Mn):
    ≤0.002 %
  • Potassium (K):
    ≤0.01 %
  • Sodium (Na):
    ≤0.05 %
  • Insoluble matter:
    ≤0.005 %
  • Nitrogen compounds (as N):
    ≤0.002 %

Specifications

Specifications

  • Catalog No:
  • ACRO111190250
  • ACRO111190010
  • ACRO111190025
  • ACRO111190251
  • Environmentally Preferable:
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Environmental certification:
  • Chem21 Solvent Selection Guide recommended by default
  • Chem21 Solvent Selection Guide recommended by default
  • Chem21 Solvent Selection Guide recommended by default
  • Chem21 Solvent Selection Guide recommended by default

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    25 mL
    Log in for availability
    Log in for availability
    1 * 25 mL
    1 * 25 mL - SFr. 25.10
    SFr. 25.10
    1 * 25 mL
       SFr. 25.10

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    ACRO111190250
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended by default
      Safer Human and Environmental Health
  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - SFr. 93.60
    SFr. 93.60
    1 * 1 L
       SFr. 93.60

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    ACRO111190010
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended by default
      Safer Human and Environmental Health
  • Glass bottle
    2,5 L
    Log in for availability
    Log in for availability
    1 * 2,5 L
    1 * 2,5 L - SFr. 185.00
    SFr. 185.00
    1 * 2,5 L
       SFr. 185.00

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    ACRO111190025
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended by default
      Safer Human and Environmental Health
  • Metal drum
    25 L
    Log in for availability
    Log in for availability
    1 * 25 L
    1 * 25 L - SFr. 957.00
    SFr. 957.00
    1 * 25 L
       SFr. 957.00

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    ACRO111190251
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended by default
      Safer Human and Environmental Health