(1R)-(+)-α-Pinene, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:7785-70-8
:
Formula:C₁₀H₁₆
MDL Number:MFCD00001346
:136.24 g/mol
Merck Index:12,07599
Boiling Pt:155 °C (1013 hPa)
Melting Pt:–62 °C
Density:0.857 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:232-087-8
UN:2368
ADR:3,III
:M-Clarity™ quality level = MQ100
Flash Pt:33 °C
Frequently Bought Together