Baths and lids, stainless steel, ST series
Supplier: GRANT INSTRUMENTS
Certificates
About this item
Premium baths which are constructed from corrosion resistant stainless steel for durability in demanding environments. Available in five sizes: 5, 12, 18, 26 and 38 litres to suit a wide variety of applications.
- All baths are supplied with mounting bridge plate. Models of 12 litres and above have a mounting plate which allows the Optima™ thermostat to be orientated on the tank in two directions - ideal for optimising bench space
- Baths of 12 litres and above feature a drain tap for easy emptying and can accommodate Grant C1G and C2G immersion coolers for operation at below ambient temperatures
- Can be used up to 200 °C
Ordering information: All baths supplied without lid. Accessory lids are recommended when working above 60 °C and should be ordered separately.
Lids
Product Information
Frequently Bought Together