


p-Xylol zur Synthese, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:106-42-3
:
Formula:C₆H₄(CH₃)₂
MDL Number:MFCD00008556
:106.17 g/mol
Merck Index:12,10214
Boiling Pt:138,3 °C (1013 hPa)
Melting Pt:13 °C
Density:1,096 g/cm³ (20 °C)
EINECS:203-396-5
UN:1307
ADR:3,III
:M-Clarity™ Qualitätsstufe = MQ200
Flash Pt:25 °C
:GHS02
:GHS07
:GHS08
Storage Temperature:Raumtemperatur
Specification Test Results
- Solubility:0.2 g/l (25 °C)
- Melting Point:13.3 °C
- Molar Mass:106.17 g/mol
- Boiling Point:138.4 °C (1013 hPa)
- Vapor Pressure:8.7 hPa (20 °C)
- Flash Point:24 °C
- Explosion Limit:1.1 - 7 %(V)
- Density:0.86 g/cm3 (20 °C)
- Ignition Point:540 °C
- Evaporation Number:13.5
Specification Test Results
Related Information
Frequently Bought Together