ContactUsLinkComponent
Hand and body lotions, Tork® Premium
Hand and body lotions, Tork® Premium
Catalog # 115-1641
Supplier:  Essity
undefined
Hand and body lotions, Tork® Premium
Catalog # 115-1641
Supplier:  Essity
Specifications
  • Description:
    Hand and body lotion, Tork® Premium, refill bottle for S2 dispenser, 475 ml
  • Pk:
    8
Specifications
About this item
Tork Premium Lotion is ideal for normal, dry and sensitive skin. It can be used as body lotion as well as hand lotion. The lotion contains canola oil, glycerin and panthenol which gives a calming, healing effect. It is also enriched with natural vitamin E (antioxidant) which restores skin properties and enhances skin barrier effect.
  • Perfect to use for frequently washed or sanitised hands
  • Perfume and colour-free, minimises the risk of allergic reactions
  • Environmental license with Nordic Ecolabel
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904