Mercury collectors
Anbieter: BURKLE
Zertifikate
Über diesen Artikel
This mercury collector enables the easy collection of spilt mercury for reusal.
- Screw cap lid with integrated foam pad to absorb mercury
- Perforated mesh insert in the container
- Pad releases mercury when squeezed against mesh insert by screwing lid onto the jar
Frequently Bought Together