Order Entry
Switzerland
ContactUsLinkComponent
 
Lidocaine hydrochloride monohydrate
Catalog # EHERC14629800
Supplier:  EHRENSTORFER
CAS Number:  
Lidocaine hydrochloride monohydrate
Catalog # EHERC14629800
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :270.8 g/mol
UN:2811
ADR:6.1,III
CAS Number:6108-05-0
Formula:C₁₄H₂₃ClN₂O·1H₂O
MDL Number:MFCD00150329