(S)-(-)-4-Methoxy-α-methylbenzylamine ≥99%, ee 98%, ChiPros®
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: (S)-1-(4-Methoxyphenyl)ethylamine, (S)-(-)-1-(4-Methoxyphenyl)ethylamine
CAS Number:41851-59-6
:
Formula:C₉H₁₃NO
MDL Number:MFCD00671660
:151.21 g/mol
Boiling Pt:65 °C (0.38 mmHg)
Melting Pt:< –20 °C
Density:1.020
Storage Temperature:Ambient
UN:2735
ADR:8,III
:GHS05
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together