Orders LinkContactUsLinkComponent
Stopcocks for separating funnels, DURAN®
Stopcocks for separating funnels, DURAN®
Supplier:   DWK Life Sciences
Stopcocks for separating funnels, DURAN®
Supplier:  DWK Life Sciences
Certificates
Stopcocks for separating funnels, DURAN®
3 Options Available Below
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear, grounded, without key.
  • Very good chemical resistance
  • Very good temperature resistance
With short inlet. For glass manipulators.
Product Family Options
Product Information
  • Spigot
    Availability
    Price
  • 12,5 NS
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - SFr. 538.00
    SFr. 538.00
    1 * 25 ST
       SFr. 538.00
  • 14,5 NS
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - SFr. 1’170.00
    SFr. 1’170.00
    1 * 50 ST
       SFr. 1’170.00
  • 18,8 NS
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - SFr. 888.00
    SFr. 888.00
    1 * 25 ST
       SFr. 888.00