Stopcocks for burettes, straight, DURAN®
Catalog # SCOT286449405
Supplier: DWK Life Sciences
Specifications
- Hole Ø:2,0 mm
- Pk:50
- Spigot:12,5 NS
Specifications
About this item
DURAN®, borosilicate glass 3.3, clear, grounded and polished, without key.
- Very good chemical resistance
- Very good temperature resistance
For glass manipulators.