Order Entry
Switzerland
ContactUsLinkComponent
 
 
 
 
Toluene ≥99.7% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
 
undefined
 
 
 
Toluene ≥99.7% (by GC), Puriss. p.a. ACS, Riedel-de Haën™

 

  • Vapor Pressure:
    26mmHg(25°C)
  • Assay (GC):
    min. 99.7%
  • Density (D20/20):
    0.865-0.869
  • Refractive index (n20/D):
    1.496-1.498
  • Non-volatilematter:
    max.0.001%
  • Water (KarlFischer):
    max.0.02%
  • Freeacid (asHCl):
    max.0.001%
  • Freealkali (asNaOH):
    max.0.001%
  • Aluminium (Al):
    max.0.00005%
  • Barium (Ba):
    max.0.00001%
  • Bismuth (Bi):
    max.0.00001%
  • Calcium (Ca):
    max.0.00005%
  • Cadmium (Cd):
    max.0.000005%
  • Cobalt (Co):
    max.0.000002%
  • Chromium (Cr):
    max.0.000002%
  • Copper (Cu):
    max.0.000002%
  • Iron (Fe):
    max.0.00001%
  • Potassium (K):
    max.0.00005%
  • Lithium (Li):
    max.0.00001%
  • Magnesium (Mg):
    max.0.00001%
  • Manganese (Mn):
    max.0.000002%
  • Molybdenum (Mo):
    max.0.00001%
  • Sodium (Na):
    max.0.00005%
  • Nickel (Ni):
    max.0.000002%
  • Lead (Pb):
    max.0.00001%
  • Strontium (Sr):
    max.0.00001%
  • Tin (Sn):
    max.0.00001%
  • Zinc (Zn):
    max.0.00001%
  • S-compounds (asS):
    max.0.003%
  • Thiophene:
    max.0.0001%
  • Reaction against H₂SO₄:
    complying
  • APHA:
    max.10
  • VaporDensity:
    3.2(vsair)

 

 

Frequently Bought Together