Order Entry
Switzerland
ContactUsLinkComponent
Toluene ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Irritant
Health
Flammable
Toluene ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Catalog # 650579-1L
CAS Number:  
undefined
Irritant
Health
Flammable
Toluene ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Catalog # 650579-1L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • IR spectrum:
    complying
  • Assay (GC):
    Min. 99.90 %
  • GC (as heptachlor epoxide):
    Max. 10 ng/l
  • Non-volatile matter:
    Max. 0.0001 %
  • Water (Karl Fischer):
    Max. 0.02 %
  • Absorbance at 284 nm:
    Max. 1.0
  • Absorbance at 300 nm:
    Max. 0.10
  • Absorbance at 325 nm:
    Max. 0.02
  • Absorbance at 350 nm:
    Max. 0.005
  • Absorbance at 400 nm:
    Max. 0.005
  • Autoignition Temperature:
    997 °F
  • Vapor Pressure:
    22 mmHg ( 20 °C)

Specification Test Results

About this item

 :
 :92.14 g/mol
Merck Index:13,09607
Boiling Pt:110.6 °C
Melting Pt:-95 °C
Melting Pt:–95 °C
Density:0.867 g/cm³ (20 °C)
Flash Pt:7.1 %(V)
Storage Temperature:Ambient
EINECS:203-625-9
UN:1294
ADR:3,II
REACH:05-2114615130-69
 :GHS07
 :GHS08
 :GHS02
CAS Number:108-88-3
Formula:C₆H₅CH₃
MDL Number:MFCD00008512