


Toluene, anhydrous (max. 0.005% H₂O) ≥99.8%, AnalaR NORMAPUR® for synthesis
28681.295
:
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
- Appearance:Clear colourless liquid
- Colour value:Max. 10 APHA
- Assay (calculated on anhydrous):Min. 99.8 %
- Residue on evaporation:Max. 0.0010 %
- Water (K.F.):Max. 0.0050 %
- Acidity or alkalinity:Max. 0.0001 meq/g
- Benzene:Max. 0.05 %
- Thiophene:0.0001 %
- Carbonisable substances:Passes test
- Al (Aluminium):Max. 0.5 ppm
- B (Boron):Max. 0.02 ppm
- Ba (Barium):Max. 0.1 ppm
- Ca (Calcium):Max. 0.5 ppm
- Cd (Cadmium):Max. 0.01 ppm
- Co (Cobalt):Max. 0.02 ppm
- Cr (Chromium):Max. 0.02 ppm
- Cu (Copper):Max. 0.01 ppm
- Fe (Iron):Max. 0.05 ppm
- Mg (Magnesium):Max. 0.1 ppm
- Mn (Manganese):Max. 0.02 ppm
- Ni (Nickel):Max. 0.01 ppm
- Pb (Lead):Max. 0.01 ppm
- Sn (Tin):Max. 0.1 ppm
- Zn (Zinc):Max. 0.01 ppm
- Total S (Sulphur):Max. 10 ppm
:
:92.14 g/mol
:13,09607
:110.6 °C (1013 hPa)
:–95 °C
:0.867 g/cm³ (20 °C)
:4 °C
:Ambient
:203-625-9
:1294
:3,II
:01-2119471310-51
:GHS02
:GHS08
:GHS07
:108-88-3
:C₆H₅CH₃
:MFCD00008512