Stopcocks for burettes, angled, DURAN®
Supplier: DWK Life Sciences
Certificates
About this item
DURAN®, borosilicate glass 3.3, with glass key.
- Very good chemical resistance
- Very good temperature resistance
- Screwthread retaining device
Frequently Bought Together