Stopcocks for burette, straight, DURAN®
Supplier: DWK Life Sciences
Certificates
About this item
DURAN®, borosilicate glass 3.3, clear, with glass or PTFE key.
- Very good chemical resistance
- Very good temperature resistance
For glass manipulators.
Frequently Bought Together