Straight valves 'GU', DURAN®
Supplier: DWK Life Sciences
Certificates
About this item
DURAN®, borosilicate glass 3.3, clear, with PTFE spindle.
- Vacuum-tight
- Precise dosing capacity due to fine thread
- Safe due to unscrewing protective device
Valves are suitable for high vacuum applications, adjustable and have heavy wall side arms.
Frequently Bought Together