Orders LinkContactUsLinkComponent
Stopcocks, Two-way
Supplier:   BURKLE
Stopcocks, Two-way
Supplier:  BURKLE
Certificates
Stopcocks, Two-way
Stopcocks, Two-way
2 Options Available Below
Product Details & Documents
HDPE, natural.
  • Resistant to acids and chemicals
  • Pressure resistant up to 0,3 bar
  • Good temperature resistance
Simple reflux nozzle with two-sided hose nozzle.
Product Family Options
Product Information
  • Hole Ø
    For tube I-Ø
    Availability
    Price
  • 7 mm
    8 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 8.00
    SFr. 8.00
    1 * 1 ST
       SFr. 8.00
  • 9 mm
    10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 12.10
    SFr. 12.10
    1 * 1 ST
       SFr. 12.10