


Toluene 99.85% for pesticide residue analysis
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:108-88-3
:
Formula:C₆H₅CH₃
MDL Number:MFCD00008512
:92.14 g/mol
Merck Index:13,09607
Boiling Pt:110.6 °C (1013 hPa)
Melting Pt:–95 °C
Density:0.866 g/cm³
Flash Pt:4 °C
Storage Temperature:Ambient
EINECS:203-625-9
UN:1294
ADR:3,II
REACH:05-2114615130-69
:GHS07
:GHS08
:GHS02
Specification Test Results
- Appearance:Clear colorless Liquid
- Water:≤0.02 % (K.F.)
- GC:≥99.80 %
- GC/ECD (as lindane):≤5 ng/l
- Residue after evaporation:≤0.0005 %
Specification Test Results
Frequently Bought Together