Examination gloves, latex, sterile, Micro-Touch® Sterile
Supplier: ANSELL MEDICAL PRODUCTS
Certificates
About this item
Micro-Touch® Sterile examination gloves are perfect for all medical applications where sterility is required and surgical gloves are not available (e.g. endoscopy, open wounds). Powder-free and with low levels of protein and residual chemicals, they minimise the risk of Type I allergies.
- Powder-free, textured fingertips
- Easy to don even with damp hands and provide excellent grip for handling instruments
- Provide a secure barrier against viruses and micro-organisms
- Interior polyacrylate coating makes the glove softer and more comfortable to wear
- Gloves are thoroughly sterilised using gamma irradiation (minimum 25kGy)
Certifications: EN 455; AQL 1,5
Packaging: Packaged in pairs with a protective double wrapping.
Specifications
- Catalog No:
- 112-1823
- 112-1824
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Disposable
- Disposable
- Powdered / powder-free:
- Powder-free
- Powder-free
- Finish:
- Textured fingertips
- Textured fingertips