Tubing clamps, stop-it
Nome del fornitore: BURKLE
Certificati
Informazioni su questo articolo
Galvanised steel and aluminium. Used to regulate or stop the flow in tubing with liquids or gases.
- Stepless flow regulation
- Heat resistant
- For hose diameters up to max. 30 mm
Frequently Bought Together