Double bossheads, cross type
: BOCHEM
Cross type double bossheads, either made of stainless steel or malleable cast iron, chromium plated.
- For rods up to 16,5 mm
- Angle: 90 + 180°
- With safety screw type 'heart' (M6)
Frequently Bought Together
Cross type double bossheads, either made of stainless steel or malleable cast iron, chromium plated.