Orders LinkContactUsLinkComponent
Bossheads, three angles
 :   BOCHEM
Bossheads, three angles
 :  BOCHEM
Bossheads, three angles
Bossheads, three angles
Three angled bossheads made of duraluminium (aluminium alloy).
  • Angle: 90 + 180 + 270°
  • Suitable for multilevel and three dimension constructions
Product Family Options
  • Material
    For rod Ø
  • Duraluminium
    9 - 13 mm
    1 * 1 ST
    1 * 1 ST - SFr. 18.30
    SFr. 18.30
    1 * 1 ST
       SFr. 18.30
  • Duraluminium
    19 - 23 mm
    1 * 1 ST
    1 * 1 ST - SFr. 32.80
    SFr. 32.80
    1 * 1 ST
       SFr. 32.80