ContactUsLinkComponent
Double bosshead, Jumbo
  BOCH5650
 :  BOCHEM
undefined
Double bosshead, Jumbo
  BOCH5650
 :  BOCHEM
  • Material:
    Aluminium
  • Pk:
    1
  • For rod Ø:
    20/30 mm
Double bossheads made of chrome plated aluminium.
  • For rods up to 20/30 mm
  • Angle: 90°
  • Thread: 10