Order Entry
Switzerland
ContactUsLinkComponent
Universal bosshead
Universal bosshead
Catalog # BOCH5334
Supplier:  BOCHEM
undefined
Universal bosshead
Catalog # BOCH5334
Supplier:  BOCHEM

Specifications

  • Material:
    Aluminium
  • Pk:
    1
  • For rod Ø:
    13 mm

Specifications

About this item

Universal aluminium bosshead with thumb screw (M8).

  • For rods up to 13 mm
  • Angle: 180 + 90°