Orders LinkContactUsLinkComponent
Boxes with Snap-On Lid, Multipurpose
Boxes with Snap-On Lid, Multipurpose
Supplier:   BURKLE
Boxes with Snap-On Lid, Multipurpose
Supplier:  BURKLE
Certificates
Boxes with Snap-On Lid, Multipurpose
Boxes with Snap-On Lid, Multipurpose
11 Options Available Below
Product Details & Documents
Box for storing, packing, taking samples, sampling and transport of pastes, granules, powders, soil samples and frozen goods (−40 °C up to briefly +95 °C).
PE, translucent.
  • Can be deep frozen
  • No crevices
  • Suitable for use with foodstuffs
Product Family Options
Round
Product Information
  • Capacity
    W×D×H
    Availability
    Price
  • BURK6210-0250
    90 mm height, 78 mm Ø
    90 mm height, 78 mm Ø
    250 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 5.40
    SFr. 5.40
    1 * 1 ST
       SFr. 5.40
  • BURK6210-0500
    127 mm height, 95 mm Ø
    127 mm height, 95 mm Ø
    500 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 6.10
    SFr. 6.10
    1 * 1 ST
       SFr. 6.10
  • BURK6210-1001
    110 mm height, 136 mm Ø
    110 mm height, 136 mm Ø
    1000 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 8.00
    SFr. 8.00
    1 * 1 ST
       SFr. 8.00
  • 216-2984
    143 mm height, 169 mm Ø
    143 mm height, 169 mm Ø
    2000 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 12.60
    SFr. 12.60
    1 * 1 ST
       SFr. 12.60
Square
Product Information
  • Capacity
    W×D×H
    Availability
    Price
  • 500 ml
    103×103×64 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 4.30
    SFr. 4.30
    1 * 1 ST
       SFr. 4.30
  • 1000 ml
    103×103×127 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 6.60
    SFr. 6.60
    1 * 1 ST
       SFr. 6.60
  • 1000 ml
    103×207×64 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 8.10
    SFr. 8.10
    1 * 1 ST
       SFr. 8.10
  • 1500 ml
    103×207×94 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 10.90
    SFr. 10.90
    1 * 1 ST
       SFr. 10.90
  • 2000 ml
    208×208×64 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 14.80
    SFr. 14.80
    1 * 1 ST
       SFr. 14.80
  • 2300 ml
    103×255×94 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 13.10
    SFr. 13.10
    1 * 1 ST
       SFr. 13.10
  • 3500 ml
    208×208×94 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 16.20
    SFr. 16.20
    1 * 1 ST
       SFr. 16.20