Wire baskets for transport
Supplier: BOCHEM
Certificates
About this item
Stainless steel 18/10.
- Three different mesh dimensions
- Stackable
- Excellent thermal and chemical resistance
These rectangular baskets are designed for transporting, storing, cleaning and sterilising laboratory utensils.
Frequently Bought Together