



2,2,4-Trimethylpentane ≥99.5% (by GC), Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:540-84-1
:
Formula:(CH₃)₂CHCH₂C(CH₃)₃
MDL Number:MFCD00008943
:114.23 g/mol
Merck Index:13,05212
Boiling Pt:99 °C (1.013 hPa)
Melting Pt:–107 °C
Density:0.69 g/cm³ (20 °C)
Flash Pt:–12 °C
EINECS:208-759-1
UN:1262
ADR:3,II
:GHS02
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Assay (GC):Min. 99.5 %
- Boiling range:98 - 100 °C
- Autoignition Temperature:745 °F
- Destillation range 98-100 °C:Min. 95 %
- Density (D 20/4):0.692 - 0.693
- Refractive index (n 20/D):1.3915 - 1.3925
- Non-volatile matter:Max. 0.001 %
- Water (Karl Fischer):Max. 0.01 %
- Free acid (as CH₃COOH):Max. 0.0005 %
- Aluminium (Al):Max. 0.00005 %
- Boron (B):Max. 0.000002 %
- Barium (Ba):Max. 0.00001 %
- Calcium (Ca):Max. 0.00005 %
- Cadmium (Cd):Max. 0.000005 %
- Cobalt (Co):Max. 0.000002 %
- Chromium (Cr):Max. 0.000002 %
- Copper (Cu):Max. 0.000002 %
- Iron (Fe):Max. 0.00001 %
- Magnesium (Mg):Max. 0.00001 %
- Manganese (Mn):Max. 0.000002 %
- Nickel (Ni):Max. 0.000002 %
- Lead (Pb):Max. 0.00001 %
- Tin (Sn):Max. 0.00001 %
- Zinc (Zn):Max. 0.00001 %
- S-compounds (as S):Max. 0.005 %
- APHA:Max. 10
Specification Test Results
Frequently Bought Together