(3-Fluoro-4-methoxyphenyl)acetic acid for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:452-14-2
:
Formula:C₉H₉FO₃
MDL Number:MFCD00060353
:M-Clarity™ quality level = MQ200
:184.17 g/mol
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Molar Mass:184.16 g/mol
Specification Test Results
Frequently Bought Together