Teclu burners, all gases
Katalognr. BOCH7430
Anbieter: BOCHEM
Technische Daten
- Max. temp.:1300 °C
- Description:Teclu multi gas burner with stopcock and air regulation
- Pk:1
- For:All gases
Technische Daten
Über diesen Artikel
Multi gas burner with stopcock works to a maximum of 1300 °C.
- With air regulation
- With anti-slip rubber coating under the base