Bunsen burners, with stopcock
Supplier: BOCHEM
Certificates
About this item
These burners reach a maximum temperature of 1300 °C and have an anti-slip rubber coating under the base.
- With air regulation sleeve
- With stopcock
- Pilot flame
Frequently Bought Together