Orders LinkContactUsLinkComponent
Beakers, low form
Beakers, low form
Supplier:   BOCHEM
Beakers, low form
Supplier:  BOCHEM
Certificates
Beakers, low form
Beakers, low form
28 Options Available Below
Product Details & Documents
Stainless steel, 18/10.
  • Good chemical resistance
  • Excellent resistance to heat
  • Smooth surfaces for fast, easy cleaning
Ideal for corrosive materials. Robust and durable.
Product Family Options
With rim
Product Information
  • Capacity
    Height
    Ext.Ø
    Availability
    Price
  • 3000 ml
    180 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 264.00
    SFr. 264.00
    1 * 1 ST
       SFr. 264.00
  • 5000 ml
    210 mm
    180 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 347.00
    SFr. 347.00
    1 * 1 ST
       SFr. 347.00
  • 500 ml
    110 mm
    80 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 119.00
    SFr. 119.00
    1 * 1 ST
       SFr. 119.00
  • 250 ml
    85 mm
    65 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 90.50
    SFr. 90.50
    1 * 1 ST
       SFr. 90.50
  • 1000 ml
    130 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 146.00
    SFr. 146.00
    1 * 1 ST
       SFr. 146.00
  • 2000 ml
    180 mm
    120 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 215.00
    SFr. 215.00
    1 * 1 ST
       SFr. 215.00
  • 100 ml
    60 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 86.70
    SFr. 86.70
    1 * 1 ST
       SFr. 86.70
With rim and spout
Product Information
  • Capacity
    Height
    Ext.Ø
    Availability
    Price
  • 2000 ml
    180 mm
    120 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 249.00
    SFr. 249.00
    1 * 1 ST
       SFr. 249.00
  • 5000 ml
    210 mm
    180 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 381.00
    SFr. 381.00
    1 * 1 ST
       SFr. 381.00
  • 1000 ml
    130 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 174.00
    SFr. 174.00
    1 * 1 ST
       SFr. 174.00
  • 3000 ml
    180 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 299.00
    SFr. 299.00
    1 * 1 ST
       SFr. 299.00
  • 100 ml
    60 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 115.00
    SFr. 115.00
    1 * 1 ST
       SFr. 115.00
  • 250 ml
    85 mm
    65 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 119.00
    SFr. 119.00
    1 * 1 ST
       SFr. 119.00
  • 500 ml
    110 mm
    80 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 147.00
    SFr. 147.00
    1 * 1 ST
       SFr. 147.00
With rim and handle
Product Information
  • Capacity
    Height
    Ext.Ø
    Availability
    Price
  • 5000 ml
    210 mm
    180 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 386.00
    SFr. 386.00
    1 * 1 ST
       SFr. 386.00
  • 100 ml
    60 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 121.00
    SFr. 121.00
    1 * 1 ST
       SFr. 121.00
  • 250 ml
    85 mm
    65 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 126.00
    SFr. 126.00
    1 * 1 ST
       SFr. 126.00
  • 500 ml
    110 mm
    80 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 153.00
    SFr. 153.00
    1 * 1 ST
       SFr. 153.00
  • 1000 ml
    130 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 180.00
    SFr. 180.00
    1 * 1 ST
       SFr. 180.00
  • 3000 ml
    180 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 306.00
    SFr. 306.00
    1 * 1 ST
       SFr. 306.00
  • 2000 ml
    180 mm
    120 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 257.00
    SFr. 257.00
    1 * 1 ST
       SFr. 257.00
With rim, spout and handle
Product Information
  • Capacity
    Height
    Ext.Ø
    Availability
    Price
  • 250 ml
    85 mm
    65 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 154.00
    SFr. 154.00
    1 * 1 ST
       SFr. 154.00
  • 500 ml
    110 mm
    80 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 182.00
    SFr. 182.00
    1 * 1 ST
       SFr. 182.00
  • 100 ml
    60 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 150.00
    SFr. 150.00
    1 * 1 ST
       SFr. 150.00
  • 1000 ml
    130 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 208.00
    SFr. 208.00
    1 * 1 ST
       SFr. 208.00
  • 2000 ml
    180 mm
    120 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 292.00
    SFr. 292.00
    1 * 1 ST
       SFr. 292.00
  • 3000 ml
    180 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 341.00
    SFr. 341.00
    1 * 1 ST
       SFr. 341.00
  • 5000 ml
    210 mm
    180 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 423.00
    SFr. 423.00
    1 * 1 ST
       SFr. 423.00