Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Phenol

Phenol

Glass bottle for solids, 1 kg
Catalog # MOLE66917197-1KG
Toxic
Corrosive
Health
Marketsource
Supplier:   Molekula
Phenol
Catalog # MOLE66917197-1KG
Supplier:  Molekula
Marketsource
CAS Number:  108-95-2
Phenol

Glass bottle for solids, 1 kg

CAS Number:  108-95-2
Price
Inquire For Price
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Phenylalcohol, Carbolic acid, Hydroxybenzene
  • CAS Number:108-95-2
  • Formula:C₆H₅OH
  • MDL Number:MFCD00002143
  • Molecular Weight:94.11 g/mol
  • Merck Index:12,07390
  • Boiling Pt:181.7 °C (1013 hPa)
  • Melting Pt:40.8 °C
  • Density:1.0722 g/cm³ (20 °C)
  • Flash Pt:81 °C
  • Storage Temperature:Ambient
  • EINECS:203-632-7
  • UN:1671
  • ADR:6.1,II
  • REACH:01-2119471329-32
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications