


Phenol 99.0-100.5%, EMPROVE® ESSENTIAL USP, Ph. Eur., ChP, JP, extra pure, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Certificates
About this item
CAS Number:108-95-2
:
Formula:C₆H₅OH
MDL Number:MFCD00002143
:94.11 g/mol
Merck Index:12,07390
Boiling Pt:181.7 °C (1013 hPa)
Melting Pt:40.8 °C
Density:1.0722 g/cm³ (20 °C)
Flash Pt:81 °C
Storage Temperature:Ambient
EINECS:203-632-7
UN:1671
ADR:6.1,II
REACH:01-2119471329-32
:GHS06
:GHS05
:GHS08
Specification Test Results
- Assay (bromatometric):99.0 - 100.5 %
- Assay (bromatometric, calc. on anhydrous substance):99.0 - 100.5 %
- Identity (IR-spectrum):passes test
- Identity (wet chemistry):passes test
- Appearance:colourless or faintly pink or faintly yellowish crystals or crystalline masses, deliquescent
- Appearance of solution (67 g/l, water):clear and not more intensely coloured than reference solution B6
- Acidic substances:passes test
- Solidification temperature:≥ 39.5 °C
- Fe (Iron):≤ 1 ppm
- Benzene (HS-GC):≤ 2 ppm
- Methanol (HS-GC):≤ 3000 ppm
- Acetone (HS-GC):≤ 2 ppm
- Anisole:≤ 10 ppm
- Cumene (HS-GC):≤ 70 ppm
- Methylisobutylketone (HS-GC):≤ 2 ppm
- Other residual solvents (ICH (Q3C)):excluded by manufacturing process
- Residue on evaporation (105 °C, 1 h):≤ 0.05 %
- Residue on evaporation (105 °C):≤ 0.05 %
- Water (according to Karl Fischer):≤ 0.5 %
Specification Test Results
Related Information
Frequently Bought Together