Order Entry
Switzerland
ContactUsLinkComponent
p-Anisidinium chloride, Sigma-Aldrich®
p-Anisidinium chloride, Sigma-Aldrich®
Catalog # 8.20103.0050
Supplier:  Merck
CAS Number:  
p-Anisidinium chloride, Sigma-Aldrich®
Catalog # 8.20103.0050
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 g
  • Seq:
    3368

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    217 - 220 °C
  • Molar Mass:
    159.62 g/mol

Specification Test Results

About this item

 :
 :159.62 g/mol
ADR:6.1,II
 :M-Clarity™ quality level = MQ200
EINECS:243-657-0
UN:3143
 :GHS06
 :GHS08
 :GHS09
Storage Temperature:Ambient
CAS Number:20265-97-8
Formula:C₇H₁₀ClNO
MDL Number:MFCD00036388