3-(Trimethylsilyl)-2-propyn-1-ol ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 3-(Trimethylsilyl)propargyl alcohol
CAS Number:5272-36-6
:
Formula:C₆H₁₂OSi
MDL Number:MFCD00002913
Boiling Pt:170…172 °C
Density:0.865
Flash Pt:66 °C (150 °F)
EINECS:226-094-5
UN:2810
ADR:6.1,III
:128.25 g/mol
:GHS06
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together