
2-Chlorocyclopentanone 97% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:694-28-0
:
Formula:C₅H₇ClO
MDL Number:MFCD00001410
:118.56 g/mol
Boiling Pt:72…74 °C (12 mmHg)
Density:1.18 g/cm³
Flash Pt:77 °C
Storage Temperature:Refrigerator
EINECS:211-769-9
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:Clear light yellow to brown Liquid (May darken on storage)
- Infrared spectrum:Conforms
- Refractive index:1.4730 to 1.4770 (20°C, 589 nm)
- GC:≥96.0 %
- Stabilizer:1 % Potassium carbonate
- UV:at 260 nm A: ≤0.1
- Color scale:≤20 APHA
Specification Test Results
Frequently Bought Together