Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

(S)-(+)-2-Hydroxyisovaleric acid 99+%

Irritant
(S)-(+)-2-Hydroxyisovaleric acid 99+%
Certificates
(S)-(+)-2-Hydroxyisovaleric acid 99+%

2 Options Available Below

Product Details & Documents
Synonyms:  (S)-2-Hydroxy-3-methylbutanoic acid, (S)-(+)-2-Hydroxy-3-methylbutanoic acid, (S)-(+)-2-Hydroxy-3-methylbutyric acid
  • CAS Number:17407-55-5
  • Formula:C₅H₁₀O₃
  • MDL Number:MFCD00066443
  • Molecular Weight:118.13 g/mol
  • Melting Pt:65…68 °C
  • Storage Temperature:Refrigerator

 

Specifications for Product Family

Specification Test Results

  • Appearance:
    White Powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    65 to 70 °C
  • GC:
    ≥99.0 %
  • Specific optical rotation:
    +15° to +19° (24 °C, 589 nm) (c=2, CHCl3)
  • Titration with NaOH:
    ≥99.0 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - SFr. 54.50
    SFr. 54.50
    1 * 1 g
       SFr. 54.50

  • Glass bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - SFr. 179.00
    SFr. 179.00
    1 * 5 g
       SFr. 179.00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.