Order Entry
Switzerland
Orders LinkContactUsLinkComponent
(S)-(+)-2-Hydroxyisovaleric acid

(S)-(+)-2-Hydroxyisovaleric acid

Marketsource
Supplier:   BACHEM BIOCHEMICA
(S)-(+)-2-Hydroxyisovaleric acid
Supplier:  BACHEM BIOCHEMICA
Marketsource
Certificates
(S)-(+)-2-Hydroxyisovaleric acid

2 Options Available Below

Product Details & Documents
Synonyms:  (S)-2-Hydroxy-3-methylbutanoic acid, (S)-(+)-2-Hydroxy-3-methylbutanoic acid, (S)-(+)-2-Hydroxy-3-methylbutyric acid
  • CAS Number:17407-55-5
  • Formula:C₅H₁₀O₃
  • MDL Number:MFCD00066443
  • Molecular Weight:118.13 g/mol
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - SFr. 112.40
    SFr. 112.40
    1 * 5 g
       SFr. 112.40

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - SFr. 449.40
    SFr. 449.40
    1 * 25 g
       SFr. 449.40

    Specifications:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.