Phenol 99+%, detached crystals
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:108-95-2
:
Formula:C₆H₅OH
MDL Number:MFCD00002143
:94.11 g/mol
Merck Index:14,7241
Boiling Pt:181 °C
Melting Pt:40…43 °C
Density:1.071
Flash Pt:79 °C (175 °F)
Storage Temperature:Ambient
EINECS:203-632-7
UN:1671
ADR:6.1,II
:GHS06
:GHS05
:GHS08
Related Information
Frequently Bought Together