Order Entry
Switzerland
ContactUsLinkComponent
Organic reference standard, 2-Naphthylamine 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Organic reference standard, 2-Naphthylamine 10 µg/ml in acetonitrile, Dr Ehrenstorfer
  EHERL10206355AL
 :  
Organic reference standard, 2-Naphthylamine 10 µg/ml in acetonitrile, Dr Ehrenstorfer
  EHERL10206355AL
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    91-59-8
  • Description:
    2-Naphthylamine 10 µg/ml in acetonitrile
  • Environmentally Preferable:
  • Formula:
    C₁₀H₉N
  • Description:
    2-Naphthylamine