Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Geraniol
Geraniol, Supelco®
Geraniol, Supelco®
Irritant
Geraniol, Supelco®
Supplier:
Merck
Certificates
Expand image
Irritant
Geraniol, Supelco®
Supplier:
Merck
Certificates
About this item
CAS Number:
106-24-1
:
Formula:
C₁₀H₁₈O
MDL Number:
MFCD00002917
:
154.25 g/mol
:
M-Clarity™ quality level = MQ100
EINECS:
203-377-1
:
GHS07
Related Information
Dokumentation
Certificates
Search by Lot Number
Dokumentation
Frequently Bought Together
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
Nitrate standard solution, 1000 mg/L NO₃<sup>-</sup> in Water (from NaNO₃) Certipur®, Supelco®
Supplier: Merck
Potassium hydrogen phthalate, Certipur® Reag. Ph. Eur., Reag. USP volumetric standard, secondary reference material for alkalimetry, Supelco®
Supplier: Merck
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Fluoride standard solution, 1,000 mg/L F- in Water (from NaF) Certipur®, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
Supplier: Merck
Potassium chloride 3 mol/l (3 N) in aqueous solution, Supelco®
Supplier: Merck
Organic reference standard, Citral, analytical standard, Supelco®
Supplier: Merck
Supelco® PESTANAL® Organic Reference Standard, Bromadiolon
Supplier: Merck
Supelco® PESTANAL® Organic Reference Standard, Pyrethrum Extract
Supplier: Merck
Supelco® PESTANAL® Organic Reference Standard, Difenacoum
Supplier: Merck