ContactUsLinkComponent
Lithium acetate
Lithium acetate
  ROTH5447.1
 :  Roth Carl
 :  
Lithium acetate
  ROTH5447.1
 :  Roth Carl
 :  
  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645
  • Environmentally Preferable:
 : Acetic acid lithium salt, Lithiumacetate
 :
 :12,05544
 :291 °C
 :1.26 g/cm³ (20 °C)
 :65.99 g/mol
 :Ambient
 :208-914-3
 :546-89-4
 :H₃CCOOLi
 :MFCD00013057