Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Tween® 20 (Polysorbate)

Tween® 20 (Polysorbate)

Glass bottle, 50 mL
Catalog # 22002.
Supplier:   Biotium
Tween® 20 (Polysorbate)
Catalog # 22002.
Supplier:  Biotium
CAS Number:  9005-64-5
Tween® 20 (Polysorbate)

Glass bottle, 50 mL

CAS Number:  9005-64-5
Price
SFr. 102.00
Log in for availability
1 * 50 mL
Product Details & Documents
Synonyms:  Polyoxyethylene sorbitan monolaurate
  • CAS Number:9005-64-5
  • Formula:C₅₈H₁₁₄O₂₆
  • MDL Number:MFCD00165986
  • Molecular Weight:1227.53 g/mol
  • Density:1.1 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:500-018-3
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.