Order Entry
Switzerland
ContactUsLinkComponent
Agarose I, powder general purpose reagent, for biotechnology low electroendosmosis (EEO)
Agarose I, powder general purpose reagent, for biotechnology low electroendosmosis (EEO)
Supplier:  G-Biosciences
Certificates
undefined
Agarose I, powder general purpose reagent, for biotechnology low electroendosmosis (EEO)
Supplier:  G-Biosciences

Specification Test Results

  • Moisture:
    ≤ 7%
  • Ash:
    0,04
  • EEO:
    0.05-0.13
  • Sulfate:
    ≤ 0.1%
  • Clarity 1.5% (NTU):
    ≤ 3
  • Gel Strength 1% (g/cm2):
    ≥ 1200
  • Gel Strength 1.5% (g/cm2):
    ≥ 2500
  • Gelling Temperature 1.5% (ºC):
    36 ± 1.5
  • Melting Temperature 1.5% (ºC):
    88 ± 1.5
  • DNase/ RNase Activity:
    None Detected
  • DNA Resolution ≥1000bp:
    Finely resolved
  • Gel background:
    Very low
  • 0.6 %:
    1,000-20,000bp
  • 0.8 %:
    500-12,000bp
  • 1.0 %:
    300-8,000bp
  • 1.2 %:
    200-6,000bp
  • 1.5 %:
    100-3,500bp
  • 2.0 %:
    50-2,000bp

Specification Test Results

Frequently Bought Together