Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

4-Nitrophenol 99%

4-Nitrophenol 99%
Certificates
4-Nitrophenol 99%

5 Options Available Below

Product Details & Documents
Synonyms:  1-Hydroxy-4-nitrobenzene, p-Nitrophenol
  • CAS Number:100-02-7
  • Formula:C₆H₅NO₃
  • MDL Number:MFCD00007331
  • Molecular Weight:139.11 g/mol
  • Merck Index:13,06654
  • Boiling Pt:279 °C (760 mmHg)
  • Melting Pt:>112 °C
  • Density:1,48 g/cm³ (20 °C)
  • Flash Pt:169 °C
  • Storage Temperature:Ambient
  • EINECS:202-811-7
  • UN:1663
  • ADR:6.1,III
Specifications for Product Family

Specification Test Results

  • Appearance:
    Yellow to brown Crystalline powder or crystals and/or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    112 to 115 °C
  • GC:
    ≥98.5 %
  • Titration with NaOH:
    ≥96.0 to ≤104.0 %
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.