
3-Nitrophenol, Millipore®
Catalog # 1.06794.0025
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:95 - 98 °C
- Molar Mass:139.11 g/mol
- Bulk Density:640 kg/m3
- Boiling Point:194 °C (93 hPa)
- Flash Point:100 °C
- Density:1.49 g/cm3 (20 °C)
- pH:6.6 - 8.6 (H2O, 20 °C)
- Ignition Point:400 °C
- Hazard Class:6.1
- Hazard Class Description:Toxic Materials
Specification Test Results
About this item
:
:139.11 g/mol
Merck Index:13,06652
Boiling Pt:271 °C (1013 hPa)
Melting Pt:94…95 °C
Density:1.485 g/cm³ (20 °C)
Flash Pt:>100 °C
Storage Temperature:Ambient
EINECS:209-073-5
UN:1663
ADR:6.1,III
:M-Clarity™ quality level = MQ200
:GHS07
CAS Number:554-84-7
Formula:C₆H₅NO₃
MDL Number:MFCD00007240