Orders LinkContactUsLinkComponent
Dissection set
Dissection set
Dissection set, 6 pcs.
Catalog # 631-1279
Supplier:   RSG
Dissection set
Catalog # 631-1279
Supplier:  RSG
Dissection set
Dissection set
Dissection set, 6 pcs.
Price
SFr. 94.90
Log in for availability
1 * 1 SET
Product Details & Documents
Dissecting set with six parts made from stainless steel, packed in a pouch.
  • 1 spoon spatula, 150 mm
  • 1 pointed tweezer, 130 mm
  • 1 powder spatula, 150/9 mm
  • 1 scalpel, 125 mm
  • 1 scissors straight, 100 mm
  • 1 spatula with blade, 150 mm
Documents
Specifications
  • Description:
    Dissection set, 6 pcs.