Nitrobenzene for synthesis, Sigma-Aldrich®
: Merck
:98-95-3
:
:C₆H₅NO₂
:MFCD00007043
:123.11 g/mol
:13,06621
:210.8 °C (1013 hPa)
:5.7 °C
:1.2037 g/cm³ (20 °C)
:202-716-0
:1662
:6.1,II
:M-Clarity™ quality level = MQ200
:88 °C
:GHS06
:GHS08
:GHS09
:Ambient
- Solubility:1.90 g/l (20 °C)
- Melting Point:6.0 °C
- Molar Mass:123.11 g/mol
- Boiling Point:211 °C (1013 hPa)
- Vapor Pressure:0.3 hPa (20 °C)
- Flash Point:88 °C
- Refractive Index:1.553 (20 °C, 589 nm)
- Explosion Limit:1.8 - 40 %(V)
- Density:1.20 g/cm3 (20 °C)
- pH:8.1 (1 g/l, H2O, 20 °C)
- Ignition Point:480 °C DIN 51794
Frequently Bought Together